Skip to Content
Merck
All Photos(1)

Key Documents

603325

Sigma-Aldrich

Methylamine-13C

99 atom % 13C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
13CH3NH2
CAS Number:
Molecular Weight:
32.05
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

99 atom % 13C

Quality Level

bp

-6.3 °C (lit.)

mp

-93 °C (lit.)

mass shift

M+1

SMILES string

[13CH3]N

InChI

1S/CH5N/c1-2/h2H2,1H3/i1+1

InChI key

BAVYZALUXZFZLV-OUBTZVSYSA-N

Packaging

All gas volumes are quoted at Standard Temperature and Pressure (STP) and not at Normal Temperature and Pressure (NTP). One STP Liter is 7.7% more gas than one NTP Liter.
Standard 500 mg quantity is packaged in a 25 mL Sure/Pac cylinder with stainless steel 1/4" male NPT valve. Nominal gas pressure at 21ºC is 29 psig for 500 mg. This pressure is slightly above atmospheric pressure. Care must be taken when extracting this product from the cylinder.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Recommended products

Stainless steel hose adapter Z14,683-8 or stainless steel body mini gas regulator Z51,354-7 is recommended.

Legal Information

SuperPak is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Gas 1A - Press. Gas Liquefied gas - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

2A - Gases

WGK

WGK 1

Flash Point(F)

-0.4 °F - closed cup

Flash Point(C)

-18 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service