Skip to Content
Merck
All Photos(1)

Key Documents

566357

Sigma-Aldrich

m-Toluenesulfonyl chloride

97%

Synonym(s):

3-Methylbenzenesulfonyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H4SO2Cl
CAS Number:
Molecular Weight:
190.65
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

refractive index

n20/D 1.5490 (lit.)

bp

252-253 °C (lit.)

density

1.314 g/mL at 25 °C (lit.)

SMILES string

Cc1cccc(c1)S(Cl)(=O)=O

InChI

1S/C7H7ClO2S/c1-6-3-2-4-7(5-6)11(8,9)10/h2-5H,1H3

InChI key

KFPMLWUKHQMEBU-UHFFFAOYSA-N

Related Categories

General description

m-Toluenesulfonyl chloride can be synthesized via chlorination of m-thiocresol in glacial acetic acid.

Application

m-Toluenesulfonyl chloride may be used in the synthesis of mesityl-m-tolyl sulfone via reaction with mesitylene.
Reactant involved in they synthesis of biologically active molecules including:
  • Small molecule ligands for the Stat3 SH2 domain
  • Isoindoline-5-propenehydroxamates for use as histone deactylase inhibitors
  • Phenyl-pyrazolylamine-based derivatives as FLT3 kinase inhibitors
  • Aryldisulfonamides with antibacterial activity
  • Quinazoline analogs for the treatment of Gaucher disease
  • 17β-HSD2 inhibitors for the treatment of osteoporosis

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

225.0 °F - closed cup

Flash Point(C)

107.2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rearrangements of sulfones to sulfinic acids via carbanion intermediates.
Truce WE and Brand WW
The Journal of Organic Chemistry, 35(6), 1828-1833 (1970)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service