Skip to Content
Merck
All Photos(1)

Key Documents

440671

Sigma-Aldrich

1,1,1,3,3,3-Hexafluoro-2-propanol-d2

≥99 atom % D, 99% (CP)

Synonym(s):

Deuterated 1,1,1,3,3,3-hexafluoro-2-propanol, Deuterated HFP, HFIP-d2, HFP-d2, Hexafluoroisopropanol-d2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CF3)2CDOD
CAS Number:
Molecular Weight:
170.05
EC Number:
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

≥99 atom % D

Quality Level

Assay

99% (CP)

form

liquid

impurities

≤0.05% water
water

refractive index

n20/D 1.275 (lit.)

bp

59 °C (lit.)

mp

−4 °C (lit.)

density

1.615 g/mL at 25 °C

mass shift

M+2

SMILES string

[2H]OC([2H])(C(F)(F)F)C(F)(F)F

InChI

1S/C3H2F6O/c4-2(5,6)1(10)3(7,8)9/h1,10H/i1D,10D

InChI key

BYEAHWXPCBROCE-AWPANEGFSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Repr. 2 - Skin Corr. 1A - STOT RE 2 Oral

Target Organs

Central nervous system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Sara La Manna et al.
Biochimie, 138, 106-115 (2017-05-10)
Interleukin-22 (IL-22) belongs to the family of IL-10 cytokines and is involved in a wide number of human diseases, including inflammatory disorders and cancer pathology. The ligand-receptor complex IL-22/IL-22R plays a key role in several pathways especially in the regulation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service