Skip to Content
Merck
All Photos(1)

Key Documents

214280

Sigma-Aldrich

2-Amino-4-methylbenzonitrile

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H3(CH3)CN
CAS Number:
Molecular Weight:
132.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

92-95 °C (lit.)

SMILES string

Cc1ccc(C#N)c(N)c1

InChI

1S/C8H8N2/c1-6-2-3-7(5-9)8(10)4-6/h2-4H,10H2,1H3

InChI key

LGNVAEIITHYWCG-UHFFFAOYSA-N

Application

2-Amino-4-methylbenzonitrile was used in the synthesis of:
  • 7-methyl-4-(phenylamino)quinazoline-2(1H)-selone
  • racemic aminoquinolines, potential acetylcholinesterase (AChE) inhibitors

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Camps et al.
Journal of medicinal chemistry, 42(17), 3227-3242 (1999-08-28)
Eleven new 12-amino-6,7,10,11-tetrahydro-7, 11-methanocycloocta[b]quinoline derivatives [tacrine (THA)-huperzine A hybrids, rac-21-31] have been synthesized as racemic mixtures and tested as acetylcholinesterase (AChE) inhibitors. For derivatives unsubstituted at the benzene ring, the highest activity was obtained for the 9-ethyl derivative rac-20, previously
Synthesis of 4-(Phenylamino) quinazoline-2 (1H)-selones and Diselenides from Isoselenocyanates: Dimroth Rearrangement of an Intermediate.
Atanassov PK, et al.
Helvetica Chimica Acta, 87(7), 1873-1877 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service