Documentos clave
509620
Syringes Fitted with Luer Lock Valve
volume 10 mL, 10MDR-VLLMA-GT, needle size (not included)
About This Item
Código UNSPSC:
41122001
Materiales
glass syringe
tamaño de la aguja
(not included)
Características
Pre-fitted with Luer Lock Valve
envase
pkg of 1 ea
fabricante / nombre comercial
SGE 008970
volumen
10 mL
Descripción general
  • SGE Syringes [volume 10 mL, needle size (not included)] fitted with Luer Lock Valve, Model: 10MDR-VLLMA-GT.
  • Fabricated using borosilicate glass, PTFE plunger tips, and KEL-F Luer cones.
  • Autoclavable syringes but the plunger removal is necessary.
  • Have an interference fit between the plunger tip and the barrel that enhances the gas sealing properties of the syringe.
  • Maximum inertness for liquid or gas samples.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)