Documentos clave
24403
SGE Syringes for Agilent/HP 7673 and 7683 Autosampler, Removable Needle
volume 0.5 μL, needle size 23 ga, needle L 42 mm, color (light orange)
About This Item
UNSPSC Code:
41122001
material
cone tip stainless steel needle (point style 1)
needle size
23 ga
description
0.5BR-HP-0.63
feature
needle type R (removable needle)
syringe type Standard syringe, removable needle
packaging
pkg of 1 ea
manufacturer/tradename
SGE 000410
needle L
42 mm
volume
0.5 μL
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)