Merck
21517
Reducer
stainless steel, 1/4 in. Swagelok, for 1/8 in. tube
About This Item
UNSPSC Code:
41115717
eCl@ss:
32035302
material
stainless steel
packaging
pkg of 1 ea
manufacturer/tradename
Swagelok® 400-R-2
fitting
for 1/8 in. tube
1/4 in. Swagelok
General description
Nuts and ferrules included.
Application
Swagelok® reducer is suitable to be placed on the ends of the tube for sampling ports used for decompositional odor analysis in the following studies:
  • Sampling tubes for graves using gas chromatography-mass spectrometry (GC-MS).
  • Sampling for Burial sites using Triple sorbent traps.
Legal Information
Swagelok is a registered trademark of Swagelok Company
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)
Lot/Batch Number
Lo sentimos, en este momento no disponemos de COAs para este producto en línea.
Visite la Librería de documentos
Arpad A Vass et al.
Journal of forensic sciences, 53(2), 384-391 (2008-03-28)
This study, conducted at the University of Tennessee's Anthropological Research Facility (ARF), lists and ranks the primary chemical constituents which define the odor of decomposition of human remains as detected at the soil surface of shallow burial sites. Triple sorbent
Arpad A Vass et al.
Journal of forensic sciences, 49(4), 760-769 (2004-08-20)
This study, conducted at the University of Tennessee's Anthropological Research Facility (ARF), describes the establishment of the Decompositional Odor Analysis (DOA) Database for the purpose of developing a man-portable, chemical sensor capable of detecting clandestine burial sites of human remains