Key Documents
21016
Supelco
Male Luer Fitting for 1/8 in. Tubing
pkg of 20 ea
About This Item
UNSPSC Code:
12190000
packaging
pkg of 20 ea
technique(s)
active air sampling: suitable
application(s)
air monitoring
environmental
industrial hygiene
General description
Male Luer Fitting for 1/8 in. Tubing is an accessory for use with TDS3 Thermal Desorption Storage Containers.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)
Lot/Batch Number
Lo sentimos, en este momento no disponemos de COAs para este producto en línea.
Visite la Librería de documentos
Los clientes también vieron
Protocolos
Simplify your thermal desorption-based terpenes sample collection in air. Learn more about their profiling, odor measurement, industrial hygiene, and environmental emissions.