Documentos clave
Z743559
WetGrip labels
white, L × W 1.28 in. × 0.5 in., roll of 1000
About This Item
UNSPSC Code:
41122700
NACRES:
NB.35
packaging
roll of 1000
manufacturer/tradename
Diversified Biotech GRRL-1000
L × W
1.28 in. × 0.5 in.
color
white
General description
Labels for Moist, Frosty or Frozen Surfaces "Grips" to wet, frost-covered and frozen surfaces Compatible with centrifuge rotors Resistant to chemicals and solvents Adheres to plastics, glass, metals and even paperboard boxes Available in white laser sheets, white direct thermal rolls, or white hand writable rolls