Key Documents
Z740294
RotoFlex® Plus bench top tube rotator rotisserie
for 5 - 7 mL tubes/7 tube capacity per rotisserie
About This Item
UNSPSC Code:
41103811
NACRES:
NB.73
packaging
pack of 2 ea
manufacturer/tradename
Cole Parmer 439737
General description
The RotoFlex will accommodate a variety of tubes from 0.5 - 50 mL by simply changing the rotisserie.
Legal Information
RotoFlex is a registered trademark of Argos Technologies, Inc.