Z740215
TLC Glass Plates
L × W 20 cm × 20 cm, PEI-cellulose, matrix, binder: no, fluorescent indicator
About This Item
Productos recomendados
material
PEI-cellulose, matrix
glass support
Quality Level
feature
binder: no
fluorescent indicator
packaging
pkg of 25 ea
manufacturer/tradename
EMD Millipore® 1.05725.0001
technique(s)
thin layer chromatography (TLC): suitable
L × W
20 cm × 20 cm
layer thickness
N/A
particle size
10-12 μm
pore size
60 Å medium pore diameter