Z370975
Dialysis tubing closures
white, size 110 mm
About This Item
Productos recomendados
material
white
size
110 mm
1 of 4
Este artículo | Z370967 | Z371092 | Z371106 |
---|---|---|---|
110 mm | 70 mm | 50 mm | 70 mm |
General description
Different colors can be used to code samples. For best results, select a closure 4-10 mm wider than the flat width of the dialysis tubing.