Documentos clave
Z332321
Bottle adapters for BRAND® Dispensers and Burettes
GL 32 thread, GL 25 bottle, ETFE
About This Item
Código UNSPSC:
41121815
NACRES:
NB.01
Materiales
ETFE
fabricante / nombre comercial
BRAND 704375
Adaptador
GL 25 bottle
GL 32 thread
Categorías relacionadas
Descripción general
For all Dispensette® models as well as seripettor® and QuikSip
Otras notas
BR704375 (India)
Información legal
BRAND is a registered trademark of BRAND GMBH + CO KG
Dispensette is a registered trademark of BRAND GMBH + CO KG
QuikSip is a trademark of BRAND GMBH + CO KG
seripettor is a registered trademark of BRAND GMBH + CO KG
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)