Key Documents
HS8171B
Spreader
L-Shape, sterile, pack of 500 (total)
Sinónimos:
Lspreader
About This Item
UNSPSC Code:
41122103
NACRES:
NB.22
sterility
sterile
packaging
bag of 10 pieces
pack of 500 (total)
L × W × thickness
146 × 38 × 4 mm
Features and Benefits
  • 10 wrap comes in zip bag
  • disposable