Documentos clave
GE28-9068-50
Hyperfilm MP
W × L 18 cm × 24 cm, pack of 50 ea (individual film envelopes)
About This Item
UNSPSC Code:
41105300
NACRES:
NB.22
sterility
non-sterile
packaging
pack of 50 ea (individual film envelopes)
manufacturer/tradename
Cytiva 28906850
W × L
18 cm × 24 cm
General description
Amersham Hyperfilm MP is an excellent multipurpose film constructed from a clear base coated on both sides with a photographic emulsion that is protected by an anti-scratch layer. The anti-scratch coating reduces the risk of visible artifacts (e.g., plastic wrap marks) and the clear colorless base results in easy reading of images, without compromising on sensitivity.
Features and Benefits
  • High sensitivity for low concentration targets.
  • Intended for radiometric or chemiluminescent applications.
  • Clear background for excellent contrast and improved band visibility.
  • Publication-quality images.
  • High resolution at an economical price.
  • Easy to use.
Storage and Stability
Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Legal Information
Hyperfilm is a trademark of Cytiva
Storage Class