Key Documents
EP6133928004
Reference Filter Set
for Eppendorf BioPhotometer® D30
About This Item
UNSPSC Code:
41104925
NACRES:
NB.24
feature
for Eppendorf BioPhotometer® D30
manufacturer/tradename
Eppendorf® 6133928004 (Global)
General description
Contains 6 filters (1 blank filter, 2 filters for checking UV/Vis wavelenght systematic error and precision, 3 filters for photometric verification)
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Eppendorf BioPhotometer is a registered trademark of Eppendorf AG