Documentos clave
EP5920729004
Plate Carrier for Eppendorf® S-4x1000 Rotor
for high-capacity buckets, pack of 2
About This Item
UNSPSC Code:
41103910
NACRES:
NB.82
packaging
pack of 2
manufacturer/tradename
Eppendorf® 5920729004 (Global)
technique(s)
centrifugation: suitable
compatibility
for use with Rotor S-4x1000
Legal Information
Eppendorf is a registered trademark of Eppendorf AG