Documentos clave
EP5427708006
Adapter for Eppendorf® F-45-18-17-Cryo Rotor
holds 1 x 1.5 mL HPLC vials, pack of 18 ea
About This Item
UNSPSC Code:
41103911
NACRES:
NB.82
feature
holds 1 x 1.5 mL HPLC vials
packaging
pack of 18 ea
manufacturer/tradename
Eppendorf® 5427708006 (Global)
technique(s)
centrifugation: suitable
compatibility
for use with Rotor F-45-18-17-Cryo
Legal Information
Eppendorf is a registered trademark of Eppendorf AG