Documentos clave
EP4960800104
Eppendorf® Bottle Thread Adapter
from GL 32 to NS 29/32, polypropylene
About This Item
UNSPSC Code:
41121815
NACRES:
NB.17
material
polypropylene
feature
from GL 32 to NS 29/32
manufacturer/tradename
Eppendorf® 4960800104
General description
Eppendorf dispensers are designed for bottle threads of 45 mm. For other bottle threads a thread adapter can be used. The diameter of the thread adapter is imprinted on the adapter. If the bottle thread is smaller than 32 mm, a second adapter must be used. If you need a thread adapter with a higer resistance to chemicals, use a PTFE/ETFE adapter.
Application
Connection of Eppendorf dispensers to bottles with neck threads different to 45 mm.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG