EP4861000740
Eppendorf® Xplorer® Plus single channel pipettor
volume 0.2-5 mL, multifunction rocker switch, purple, for pipette tips 5 mL
About This Item
Código UNSPSC:
41121500
NACRES:
NB.01
Productos recomendados
Características
for pipette tips 5 mL
multifunction rocker switch
envase
case of 1 ea
fabricante / nombre comercial
Eppendorf® 4861000740
volumen
0.2-5 mL
color
purple
Categorías relacionadas
Descripción general
- Multi-functional rocker with up is up and down is down functionality
- Selection dial for quick and easy selection of functions
- Self-explanatory color display without submenus and with ergonomic angle for optimal readability in every position
- Spring-loaded tip cone for all pipettes up to 1,000 μL for minimal tip attachment forces helps to reduce stress
- Powerful rechargeable battery for up to 8 hours work without the need for recharging
- Separate power socket: Work can continue while charging
- Adjustment to external conditions (e.g. geographic height, liquids with high density)
- Lower part of pipette can be autoclaved to ensure decontamination
- 9 different languages: Chinese, Dutch, English, French, German, Italian, Japanese, Portuguese and Spanish
- Xplorer plus only: Individual programming, fixed volume settings, password protection, on/off switch, additional applications, display of service intervals
Información legal
Eppendorf is a registered trademark of Eppendorf AG
Xplorer is a registered trademark of Eppendorf AG
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)