Documentos clave
CLS6406125FO
PYREX® replacement Squibb separatory funnel w/o valve and stopper
Sinónimos:
equipment replacement accessory
About This Item
UNSPSC Code:
41121809
packaging
pack of 4 ea
manufacturer/tradename
Corning 6406-125FO
capacity
125 mL
volume
125 mL
General description
These economy Squibb separatory funnel replacements do not have a Polytetrafluorethylene (PTFE) valve assembly or polyethylene stopper.
Legal Information
PYREX is a registered trademark of Corning, Inc.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)