Documentos clave
Vero76
85020205, African green monkey kidney, Epithelial
About This Item
UNSPSC Code:
41106514
Nombre del producto
Vero76, from African green monkey kidney, 85020205, Convention on the International Trade in Endangered Species of Wild Fauna and Flora (CITES)
biological source
African green monkey kidney
growth mode
Adherent
karyotype
2n = 60
morphology
Epithelial
receptors
Not specified
Cell Line Origin
Monkey African Green kidney
Cell Line Description
Vero76 was derived from the original parent Vero line (Sigma catalog no. 84113001) and exhibits a lower saturation density than the parent line. The cells are susceptible to human haemorrhagic fever viruses.
Culture Medium
DMEM (D6546) + 2 mM L-Glutamine (G7513) + 10% FBS / FCS (F2442).
Subculture Routine
Split sub-confluent cultures (70-80%) 1:3 to 1:10, i.e., seeding at 1-3x10,000 cells/cm2 using 0.25% trypsin or trypsin/EDTA; 5% CO2; 37 °C.
Other Notes
Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)