AXYPCR0108LPC
Axygen low profile 0.1 mL thin wall PCR strip tubes
clear
About This Item
Productos recomendados
material
clear
polypropylene
sterility
non-sterile
packaging
pack of 125 ea
case of 1250 ea
manufacturer/tradename
Corning PCR-0108-LP-C
working volume
5-125 μL
capacity
0.1 mL
suitability
suitable for PCR