Key Documents
AXYAP850
AxyPet® pipette
8 channel, autoclavable, size 5-50 μL, pkg of 1 ea
Sinónimos:
pipet
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
feature
8-channel
electronic: no
packaging
pkg of 1 ea
manufacturer/tradename
Axygen Inc AP-8-50
Corning
size
5-50 μL
Legal Information
AxyPet is a registered trademark of Axygen, Inc.