Merck
915
Sigma-Aldrich
Citrate Solution
pH 3.6±0.1 (25 °C), 27 mM
About This Item
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.47
form
solution
Quality Level
IVD
for in vitro diagnostic use
concentration
27 mM
technique(s)
titration: suitable
pH
3.6±0.1 (25 °C)
application(s)
hematology
histology
shipped in
wet ice
storage temp.
2-8°C
SMILES string
[Na+].[Na+].[Na+].OC(CC([O-])=O)(CC([O-])=O)C([O-])=O
InChI
1S/C6H8O7.3Na/c7-3(8)1-6(13,5(11)12)2-4(9)10;;;/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);;;/q;3*+1/p-3
Inchi Key
HRXKRNGNAMMEHJ-UHFFFAOYSA-K
Application
Used as a kit component for the 91A and 91C procedures for Esterase.
Components
Citric Acid, 18 mmol/l, sodium citrate, 9 mmol/L, sodium chloride, 12 mmol/L with surfactant. pH 3.6 +/- 0.1 at 25 °C.
Storage Class
12 - Non Combustible Liquids
wgk_germany
nwg
flash_point_f
Not applicable
flash_point_c
Visite la Librería de documentos
Nuoya Yin et al.
Chemosphere, 204, 109-118 (2018-04-15)
F-53B, as an alternative to the persistent organic pollutant perfluorooctane sulfonate (PFOS), is amply used in the electric plating industry. F-53B and PFOS have similar physicochemical, biochemical and physiological properties, due to the similarity in their chemical structure. Thus, they
Alessio Innocenti et al.
Bioorganic & medicinal chemistry letters, 15(3), 573-578 (2005-01-25)
A detailed inhibition study of five carbonic anhydrase (CA, EC 4.2.1.1) isozymes with carboxylates including aliphatic (formate, acetate), dicarboxylic (oxalate, malonate), hydroxy/keto acids (l-lactate, l-malate, pyruvate), tricarboxylic (citrate), or aromatic (benzoate, tetrafluorobenzoate) representatives, some of which are important intermediates in
Alessio Innocenti et al.
Bioorganic & medicinal chemistry, 17(7), 2654-2657 (2009-03-20)
The inhibition of the beta-carbonic anhydrases (CAs, EC 4.2.1.1) from the pathogenic fungi Cryptococcus neoformans (Can2) and Candida albicans (Nce103) with carboxylates such as the C1-C5 aliphatic carboxylates, oxalate, malonate, maleate, malate, pyruvate, lactate, citrate and some benzoates has been