Documentos clave
1.51419
Supelco
manu-CART® "10"-II cartridge holder for LiChroCART® 10 mm i.d. HPLC-cartridges
About This Item
UNSPSC Code:
41115709
technique(s)
HPLC: suitable
Quality Level
storage temp.
no temp limit
Categorías relacionadas
General description
manu-CART "10" II cartridge holder for 10 mm i.d. LiChroCART cartridges.
Analysis Note
Extent of testing (visual test): passes test
Legal Information
LICHROCART is a trademark of Merck KGaA, Darmstadt, Germany
MANU-CART is a trademark of Merck KGaA, Darmstadt, Germany