Key Documents
Z742456
Block for Multi-Therm shaker
Holds 1 x deep well mircoplate, max. height 54 mm
About This Item
UNSPSC Code:
41102404
NACRES:
NB.73
feature
Holds 1 x deep well mircoplate, max. height 54 mm
packaging
pack of 1 ea
manufacturer/tradename
Benchmark Scientific H5000-DWMP
General description
For use with Benchmark Multi-Therm Shaker
Legal Information
Multi-Therm is a trademark of Benchmark Scientific, Inc.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)