Z740714
Aldrich® Essentials boiling flask, round-bottom, ST/NS 24/29
capacity 100 mL
About This Item
Productos recomendados
material
borosilicate glass flask
round bottom
single-neck
packaging
pkg of 1 ea
capacity
100 mL
General description
Clear borosilicate glass, round bottomed boiling flasks with outer ST ground joint neck. White marking spot.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)