Merck
Z568082
PureSolv Micro replacement purification column
activated molecular sieves
About This Item
UNSPSC Code:
40161600
NACRES:
NB.85
manufacturer/tradename
Innovative Technologies
General description
Sieve type 5A
Legal Information
Pure Solv is a trademark of Inert Corp.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)