Z558990
Sigma-Aldrich® anti-static disposable unisex lab coat
light blue, size XL
About This Item
Productos recomendados
size
XL
availability
not available in EU
color
light blue
General description
DL4260 Disposable Lab Coats. These tough, lightweight, full-length lab coats are splash/spray resistant and offer superior comfort/breathability, and are anti-static treated.
Details:
Details:
- Material: SMS
- Weight: 30 grams
- Collar: knit
- Pockets: 3
- Cuffs, Heavy Duty Snaps, Latex Free: yes
Legal Information
SIGMA-ALDRICH is a registered trademark of Merck KGaA, Darmstadt, Germany
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)