Key Documents
Z549592
Sigma-Aldrich
Stopcock Adapter
glass side-port stopcock, joint: ST/NS 29/32
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
material
glass side-port stopcock
joint
joint: ST/NS29/32
General description
Glass and PTFE stopcocks have a 2 mm bore and accommodate 1/4 in. I.D. tubing. Can be used in vacuum or atmospheric applications.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)