Documentos clave
Z531154
Sigma-Aldrich
Aldrich® coiled condenser
unjacketed, L 300 mm, joint: ST/NS 29/32
About This Item
Código UNSPSC:
41121800
NACRES:
NB.43
Junta
joint: ST/NS29/32
Longitud
300 mm
Categorías relacionadas
Descripción general
The coiled condenser circulates coolant through the coil. The jacketed version provides additional cooling capacity by allowing vapors to condense on the inner wall of the jacket.
Información legal
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)