Documentos clave
Z530395
Sigma-Aldrich
Aldrich® short-path distillation head
thermometer joint: ST/NS 10/18, Joints: ST/NS 19/22 (2)
About This Item
UNSPSC Code:
41104812
NACRES:
NB.43
joint
thermometer joint: ST/NS10/18
Joints: ST/NS19/22 (2)
immersion depth
50 mm
Categorías relacionadas
General description
Vigreux indents prevent bumping of fine or lightweight column packing into the condenser or fraction cutter when column is flooded.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)