Documentos clave
Z529532
Coors porcelain pestle
for Z529508
About This Item
UNSPSC Code:
41121800
NACRES:
NB.35
material
porcelain
packaging
pack of 1 ea
manufacturer/tradename
Coorstek 66329
pestle L
262 mm
General description
Oversized pestle glazed to grinding surface. Order mortar separately.
Application
The Coors porcelain pestle is suitable to use for crushing, grinding, and mixing.
Legal Information
Coors is a trademark of Coorstek, Inc.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)