Z515876
Aldrich® solvent storage flask
capacity 50 mL, joint: ST/NS 14/20
Sinónimos:
bottom flask, flask round, round flask
About This Item
Productos recomendados
material
borosilicate glass
round bottom
single-neck
joint
joint: ST/NS14/20
sterility
non-sterile
feature
closure type Standard Taper Joint
packaging
pack of 1
manufacturer/tradename
Sigma-Aldrich
capacity
50 mL
General description
Legal Information
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)