Z287253
Aldrich® low-pressure basic three station manifold
hose outlet I.D. 1/4 in.
About This Item
Productos recomendados
hose outlet I.D.
1/4 in.
General description
This system offers complete manifold installation flexibility.
Made of brass with 1/4 in. NPTM inlet. Consists of three stations, each with a needle valve and 1/4 in. I.D. hose barb outlet.
As increased manifold capacity is needed, simply order the required parts and install them on-site (refer to ′Related Products′ tab).
Made of brass with 1/4 in. NPTM inlet. Consists of three stations, each with a needle valve and 1/4 in. I.D. hose barb outlet.
As increased manifold capacity is needed, simply order the required parts and install them on-site (refer to ′Related Products′ tab).
Caution
Do not exceed 200 psi inlet gas pressure.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
For use with
Referencia del producto
Descripción
Precios
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)