Key Documents
Z275298
Sigma-Aldrich
Cannon-Fenske viscometer tube
size 75
About This Item
UNSPSC Code:
41111700
NACRES:
PB.36
size
75
description
constant ~ 0.008 cSt/s
manufacturer/tradename
Ace Glass 798837
viscosity measuring range
1.6-8 cSt
General description
Obtain kinematic viscosities using tubes designed for ASTM Method D445 and D2515. Viscosities can be measured with a precision of ±0.2% quickly and easily.
Requires approximately 7 mL of sample. Certificate of Calibration supplied.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)