Key Documents
Z247731
Coors high-alumina crucible cover
diam. 48 mm
About This Item
UNSPSC Code:
41122200
NACRES:
NB.43
packaging
pack of 1 ea
manufacturer/tradename
Coorstek 65611
diam.
48 mm
General description
Covers may not fit exactly on crucibles due to unavoidable wall thickness variations within the crucible.
For Z247634
Legal Information
Coors is a trademark of Coorstek, Inc.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)