Key Documents
Z224863
Sigma-Aldrich
Replacement stopper for Aldrich® combustion apparatus
Replacement stopper
About This Item
UNSPSC Code:
41122500
NACRES:
NB.43
material
borosilicate glass
sterility
non-sterile
packaging
pack of 1
manufacturer/tradename
Sigma-Aldrich
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)