Key Documents
Z216011
Solid PTFE stopper
joint: ST/NS 34/35
Sinónimos:
flask stopper, lab stopper, stoppers
About This Item
UNSPSC Code:
41100000
NACRES:
NB.35
joint
joint: ST/NS34/35
manufacturer/tradename
Cowie (010.234)
General description
Machined from solid PTFE with trapezoidal seals for medium to high vacuum performance up to 280°C (540°F).
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)