Key Documents
Z215473
Rigid PTFE sleeve
joint: ST/NS 14/23
Sinónimos:
ptfe sleeves
About This Item
UNSPSC Code:
41100000
NACRES:
NB.35
joint
joint: ST/NS14/23
manufacturer/tradename
Cowie
General description
Robust, PTFE sleeve with substantial grip-ring for repeated use and ease of handling. Transverse trapezoidal sealing rings provide grease-free, high-vacuum performance at temperatures up to 280°C.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)