Key Documents
Z203017
Supelco
Flow controller for flash-chromatography assembly with threaded joints
joint: ST/NS 29/32
About This Item
UNSPSC Code:
12000000
NACRES:
SB.52
joint
joint: ST/NS29/32
grade
for chromatography
technique(s)
LPLC: suitable
Linkage
See the Threaded Ground Glass Tutorial to see how threaded joints work.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)