Key Documents
Z202622
Sigma-Aldrich
Aldrich® Luer lock adapter
male joint: ST/NS 24/40 to male Luer lock
About This Item
UNSPSC Code:
41121815
NACRES:
NB.35
joint
male joint: ST/NS24/40 to male Luer lock
General description
PTFE needle valve and 11/2 in. (38 mm) drip tip.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)