Key Documents
Z189707
Sigma-Aldrich
Sleeves for dry box glove and sleeve assembly
straight sleeves, neoprene, port size 6 in.
About This Item
UNSPSC Code:
46181504
NACRES:
NB.35
material
neoprene
sterility
non-sterile
feature
latex free
powder-free
reusable
packaging
pack of 1 pair
manufacturer/tradename
Sigma-Aldrich
port size
6 in.
Packaging
Package includes 2 sleeves, wrist rings, and springs.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)