Merck
Todas las fotos(6)
Key Documents
Z168874
Scissor-handle forceps
curved, Kelly hemostatic, L 6 1/4 in.
About This Item
UNSPSC Code:
41122405
NACRES:
NB.35
label
C
packaging
pack of 1 ea
L
6 1/4 in.
General description
with box-lock joint and serrated jaws