Key Documents
Z163988
Sigma-Aldrich
Gravity column
Long
About This Item
UNSPSC Code:
23151816
NACRES:
NB.51
joint
joint ST/NS24/40
L × I.D. × O.D.
70 cm × 24 mm × 28 mm
Application
Gravity column has been used in the separation of Ixora flower petal extracts by liquid column chromatography (LCC).
Features and Benefits
  • Provided with a 4mm polytetrafluoroethylene (PTFE) stopcock
  • Has no frit and uses a glass-wool plug and sand
  • Use along with coarse silica gel (e.g., Cat. No. 60744) for fast and general-purpose purification.
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)