Key Documents
Z153109
Sigma-Aldrich
Aldrich® jacketed round-bottom flasks
capacity 1,000 mL
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
material
borosilicate glass
round bottom flask
three-neck
sterility
non-sterile
feature
closure type Standard Taper Joint
packaging
pack of 1
manufacturer/tradename
Sigma-Aldrich
capacity
1,000 mL
General description
24/40 joint, with two hose connections. Multifunctional-can be water cooled or heated with hot water or steam.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)