Key Documents
Z125784
Sigma-Aldrich
Ace three-neck round-bottom flasks
angled, capacity 5,000 mL, Joints: ST/NS 24/40 (3)
Sinónimos:
bottom flask, flask round, round flask
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
material
borosilicate glass
round bottom flask
three angled neck
joint
Joints: ST/NS24/40 (3)
sterility
non-sterile
feature
closure type Standard Taper joint
packaging
pack of 1
manufacturer/tradename
Ace Glass 6948-58
capacity
5,000 mL
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)