Key Documents
Z121525
Sigma-Aldrich
Aldrich® cold-trap with ball joint
joint: SJ 75/50
About This Item
UNSPSC Code:
41104812
NACRES:
NB.43
joint
joint: SJ75/50
General description
Trap 7 cm I.D. × 30 cm Length, overall 75 mm O.D. × 470 mm Length, 20 mm O.D. side tubes. Includes pinch clamp.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
replacement part
Referencia del producto
Descripción
Precios
Elija entre una de las versiones más recientes:
Certificados de análisis (COA)