Z118796
®Pulgas agitadoras magnéticas Spinbar
yellow, diam. 2 mm, L 5 mm
About This Item
UNSPSC Code:
41103806
NACRES:
NB.72
Productos recomendados
material
PTFE-coated
cylindrical PTFE-coated bar
feature
ALNICO V coated magnet
packaging
pack of 3 ea
manufacturer/tradename
Bel-Art F37121-0011
L
5 mm
diam.
2 mm
color
yellow
General description
- Spinbar® magnetic stirring fleas are designed for use in tubes, vials, and gradient markers.
- Ideal for mixing of small volume samples in environmental testing and life science applications.
- Alnico magnet encapsulated in durable and inert FDA grade PTFE (polytetrafluoroethylene) coating.
- Broad chemical resistance for use in a wide variety of applications.
- Autoclavable.
Legal Information
Spinbar is a registered trademark of SP Scienceware, a division of SP
Teflon is a registered trademark of E. I. du Pont de Nemours and Company